Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® Single-Channel Digital Microdispensers, Variable Volume

VWR® Single-Channel Digital Microdispensers, Variable Volume

10 - 50 µl, 0.1 µl, Green
  53506-143
VWR® Single-Channel Digital Microdispensers, Variable Volume
  53506-143
VWR® Single-Channel Digital Microdispensers, Variable Volume
VWR® Single-Channel Digital Microdispensers, Variable Volume

10 - 50 µl, 0.1 µl, Green

 
$1,149.33
   
Each
 

This adjustable dispenser features a digital readout, and a positive displacement piston ensures complete recovery of the sample.

  • Accurate dual-calibration system
  • Easy to set and easy to use
  • Positive displacement pipetting of any fluid
  • Adjustable
  • Precision-molded plastic or glass capillary bores

A unique dual-calibration system provides an accuracy and reproducibility of better than ±1%.

Capillary bores are made of glass except for the 100 to 1000 µl volume and 1000 µl volume models, which are made of precision-molded plastic. Users can set whole or fractional microliter volumes.

A cutout in the barrel of the unit permits a visual check for proper insertion of the capillary bore. A calibration mark on each bore allows for a visual check for complete delivery, and a color-coded mark on the bore lines up with the stainless steel plunger when the dispenser is set at full capacity.

Ordering information: The 10 µl unit comes with replacement bores and an Allen wrench. The larger dispensers are supplied with an accessory kit that includes replacement bores, an Allen wrench, replacement fluoropolymer resin plunger tips, a tip-replacement block, tip resizing tools, and a plunger replacement gauge.

Services: "Premium Pre-Calibrated Cobra® Pipettors include ISO 17025 NIST Traceble Calibration Certificates, including 10 measurments at 3 volumes. All calibrations are conducted under an ISO/IEC 17025 accredited laboratory. Certificate includes all data as found and as left (and uncertainties).

VWRCatalyst™ Services provides convenient and efficient recalibration services for pipettors, with an industry leading 3 business day average turn-around time from receipt of the instrument. Services are performed by a VWR® selected ISO/IEC 17025, NVLAP accredited supplier (NVLAP Lab code 105013-0), with calibration certificate traceable to NIST. Recalibration, ISO 17025 certification, and shipping fees are all included in the catalog number cost. Three different levels of calibration can be selected based on specific needs.
Customers will receive a specially designed protective box to ship the instrument to a certified calibration laboratory, along with a work order form and a pre-paid return shipping label. Upon receipt at the laboratory, the instrument will be inspected for any defects and the performance of the instrument will be compared to certified standards. After recalibration, the calibration report and instrument will be carefully repacked and returned with standard ground shipping.
In addition, VWRCATALYST™ can provide on-site pipettor calibration service. Please call 888-793-2300 or email VWR[email protected] for further details."

Documents

 
  • Volume:
    10 - 50 µl
  • Increments:
    0.1 µl
  • Color code:
    Green
Accessories

 

  •  
    Description
    For use with
     
     
  • 53508-400
    Replacement Capillary Bores, Orange
    10 µL microdispensers
       
    Pack of 100 - $49.48

 

  •  
    Description
    Volume
     
     
  • 53506-902
    Replacement Part Kit
    10 µl
       
    Each - $39.98

 

  •  
    Description
    Volume
     
     
  • 53506-904
    Replacement Part Kit
    25 µl
       
    Each - $41.34

 

  •  
    Description
    Volume
     
     
  • 53506-906
    Replacement Part Kit
    50 µl
       
    Each - $41.34

 

  •  
    Description
    Volume
     
     
  • 53506-908
    Replacement Part Kit
    100 µL
       
    Each - $39.21

 

  •  
    Description
    For use with
     
     
  • 53508-375
    Replacement Capillary Bores, White
    1, 2, 3, 4, 5 µL microdispensers
       
    Pack of 100 - $48.42

 

  •  
    Description
    For use with
     
     
  • 53508-386
    Replacement Capillary Bores, Orange
    3.33, 6.66 µL microdispensers
       
    Pack of 100 - $48.73

 

  •  
    Description
    For use with
     
     
  • 53498-162
    Replacement Fluoropolymer Resin-Coated Tips
    1000 µL microdispensers
       
    Pack of 3 - $19.17

 

  •  
    Description
    For use with
     
     
  • 53508-535
    Replacement Capillary Bores, Black
    20 µL microdispensers
       
    Pack of 100 - $48.73

 

  •  
    Description
    For use with
     
     
  • 53508-433
    Replacement Capillary Bores, 2 White
    25 µL microdispensers
       
    Pack of 100 - $49.36

 

  •  
    Description
    For use with
     
     
  • 53508-590
    Replacement Capillary Bores, Black
    250 µL microdispensers
       
    Pack of 100 - $48.87

 

  •  
    Description
    For use with
     
     
  • 53508-466
    Replacement Capillary Bores, Green
    50 µL microdispensers
       
    Pack of 100 - $49.36

 

  •  
    Description
    For use with
     
     
  • 53508-499
    Replacement Capillary Bores, Blue
    100 µL microdispensers
       
    Pack of 100 - $49.12

 

  •  
    Description
    For use with
     
     
  • 53508-660
    Replacement Capillary Bores, Clear
    1000 µL microdispensers
       
    Pack of 100 - $55.87