Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

PYREX® Serological Pipettes, Corning

Pipet, sterile serological Pyrex®, 2 ml, 0.2 ml, 0.01 ml, Individually Packed
Catalog # 53284-347
Supplier:   Corning
 :  7077-2N
PYREX® Serological Pipettes, Corning
Catalog # 53284-347
Supplier:  Corning
PYREX® Serological Pipettes, Corning
PYREX® Serological Pipettes, Corning

Pipet, sterile serological Pyrex®, 2 ml, 0.2 ml, 0.01 ml, Individually Packed

Price
$998.00
Log in for availability
Case of 720
Product Details & Documents

These sterile disposable 1 ml PYREX® serological pipettes, calibrated (TD) with blow-out, offer long, slender, tapered tips to make pipetting go faster and easier.

  • All pipettes have negative graduations
  • Fire-polished tips for burr-free, even flow rates

Easy-open Steri-View™ wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size.

 : Colors conform to I.S.O. standards.

 : Non sterile pipettes available in bulk package; sterile pipettes available in individual paper-plastic wrappers or as Multi-Pack in plastic bags.

Specifications
  • Capacity:
    2 ml
  • Description:
    Pipet, sterile serological Pyrex®
  • Packed:
    Individually Packed
  • Subdivisions:
    0.01 ml
  • Negative graduations:
    0.2 ml