Generators for Polytron® PT Models, Kinematica
PT-DA 07 / 2EC-E107, Saw Tooth, 0.10 - 5 ml, 7 mm, 7×107 mm, PT 1200 E, PT 1300 D, PT 2500 E
Catalog # 47751-628
Supplier: Kinematica
Supplier #: 11030031
Jump to:
Product Details & Documents
Specifications
- Ø:7 mm
- Description:PT-DA 07 / 2EC-E107, Saw Tooth
- Diameter ×L:7×107 mm
- For use with:PT 1200 E, PT 1300 D, PT 2500 E
- Sample volume:0.10 - 5 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



