Caliper Electrodes, BTX
Brass Caliper Electrode, 1.0 x 1.0 cm, 1 - 130 mm
Catalog # 47727-352
Supplier: Harvard Apparatus
Supplier #: 45-0101
Jump to:
Product Details & Documents
In vivo-style, reusable electrodes are suitable for a variety of applications, including gene and transdermal drug delivery
Electrodes are compatible with electroporation systems 58018-004 and 58017-932. Electrodes complement Tweezertrodes™ 22221-518 and -520; Genetrodes® 47727-356 and -358; and 2-Needle Array™ electrode kits 22221-522 and -524.
Calipers may be cleaned with a mild detergent and sterilized with ethanol or ethylene oxide.
Each model consists of a caliper and a pair of end plates. End plates are adjusted using the black roller mounted on the caliper. Use brass caliper for smaller animals with a smaller surface area. Use stainless steel calipers for slightly larger surface areas.
Specifications
- Description:Brass Caliper Electrode
- Dimensions:1.0 x 1.0 cm
- Gap size:1 - 130 mm
- Cat. No.:47727-352
- Pulse Length Range:1µs–99ms
- Voltage Range:0 - 500 V



