VWR® Basic SMS Frocks
Product Details & Documents
Basic Protection SMS Frocks are designed to provide lightweight, breathable coverage for non-hazardous environments. Made from SMS (Spunbond-Meltblown-Spunbond) material, they offer a dependable barrier against dust, dirt, and light liquid splashes while ensuring comfort during extended wear.
Spunbond-Meltblown-Spunbond construction for lightweight durability and breathability
Snap-front closure is easy to wear and remove while maintaining a secure fit
Elastic cuffs help to keep sleeves in place
Full-length coverage provides protection for clothing and skin in controlled environments
Reliable barrier protection; shields against common workplace contaminants
Clean, professional design suitable for labs and production areas
Affordable option for environments requiring basic protection
The Basic Protection SMS Frock is engineered for environments where comfort and essential coverage are key. Constructed from SMS (Spunbond-Meltblown-Spunbond) material, this garment combines durability with breathability, making it suitable for extended wear in controlled settings. The SMS fabric’s multi-layer design provides a dependable barrier against dust, dirt, and light liquid splashes, ensuring protection without sacrificing mobility.
Featuring a snap-front closure for easy donning and doffing, along with elastic cuffs for a secure fit, the frock maintains a professional appearance while enhancing user safety. The lightweight structure reduces heat buildup, promoting comfort during long shifts. Ideal for laboratories, cleanrooms, and production areas, this frock delivers practical protection for non-hazardous applications where compliance and convenience matter.
Certifications: Frocks are rigorously tested and manufactured in an ISO certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Packaging: Frocks are packaged 30 per case.
Documents
Certificates
Search by Lot NumberVWR Basic Protection SMS Frocks
Datasheet
VWR Protection Apparel
Product Literature
Specifications
- Clothing Size:M
- Color:White
- Length:42¹/₂"
- Chest size:22³/₄"
- Sleeve:34¹/₂"
- Tensile Strength:13 lbs./in. (warp), 16.1 lbs./in. (filling)
- Air Permeability:95.3 cu. ft./min./sq. ft.
- Bacterial Filtration Efficiency:90%
- Sleeve or leg length:Long
- Tearing Strength:3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
- Gender:Unisex
- Closure type:With Press-Stud Fastening
- Weight:50.45 g/m²
- H₂O Resistance:16.7 G
- Cat. No.:414004-389
- Water Vapor Transmission Rate:965 g/m²/24 h
- Cleanroom Class:10000/ISO 7
- Garment application:Cleanroom|Industrial|Laboratory
- Pockets:With Pockets
- Fabric:Spunbound polypropylene
- Particle shedding (Helmke Drum):Level II
Complementary Products
Product Information
- Catalog #DescriptionSizeAvailabilityPrice
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



