Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® Basic SMS Frocks

VWR® Basic SMS Frocks

White, XL, 46", 24³/₄", 36¹/₂"
Catalog # 414004-386
Supplier:   VWR International
VWR® Basic SMS Frocks
Catalog # 414004-386
Supplier:  VWR International
VWR® Basic SMS Frocks
VWR® Basic SMS Frocks

White, XL, 46", 24³/₄", 36¹/₂"

Price
$366.05
Log in for availability
Case of 30 Pallet of 720
Product Details & Documents

Basic Protection SMS Frocks are designed to provide lightweight, breathable coverage for non-hazardous environments. Made from SMS (Spunbond-Meltblown-Spunbond) material, they offer a dependable barrier against dust, dirt, and light liquid splashes while ensuring comfort during extended wear.

  • Spunbond-Meltblown-Spunbond construction for lightweight durability and breathability

    Snap-front closure is easy to wear and remove while maintaining a secure fit

    Elastic cuffs help to keep sleeves in place

    Full-length coverage provides protection for clothing and skin in controlled environments

    Reliable barrier protection; shields against common workplace contaminants

    Clean, professional design suitable for labs and production areas

    Affordable option for environments requiring basic protection

The Basic Protection SMS Frock is engineered for environments where comfort and essential coverage are key. Constructed from SMS (Spunbond-Meltblown-Spunbond) material, this garment combines durability with breathability, making it suitable for extended wear in controlled settings. The SMS fabric’s multi-layer design provides a dependable barrier against dust, dirt, and light liquid splashes, ensuring protection without sacrificing mobility.

Featuring a snap-front closure for easy donning and doffing, along with elastic cuffs for a secure fit, the frock maintains a professional appearance while enhancing user safety. The lightweight structure reduces heat buildup, promoting comfort during long shifts. Ideal for laboratories, cleanrooms, and production areas, this frock delivers practical protection for non-hazardous applications where compliance and convenience matter.

Certifications: Frocks are rigorously tested and manufactured in an ISO certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.

Packaging: Frocks are packaged 30 per case.

Specifications
  • Clothing Size:
    XL
  • Color:
    White
  • Length:
    46"
  • Chest size:
    24³/₄"
  • Sleeve:
    36¹/₂"
  • Tensile Strength:
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • Air Permeability:
    95.3 cu. ft./min./sq. ft.
  • Bacterial Filtration Efficiency:
    90%
  • Sleeve or leg length:
    Long
  • Tearing Strength:
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • Gender:
    Unisex
  • Closure type:
    With Press-Stud Fastening
  • Weight:
    50.45 g/m²
  • H₂O Resistance:
    16.7 G
  • Cat. No.:
    414004-386
  • Water Vapor Transmission Rate:
    965 g/m²/24 h
  • Cleanroom Class:
    10000/ISO 7
  • Garment application:
    Cleanroom|Industrial|Laboratory
  • Pockets:
    With Pockets
  • Fabric:
    Spunbound polypropylene
  • Particle shedding (Helmke Drum):
    Level II
Complementary Products

Product Information

  • Catalog #
    Description
    Size
    Availability
    Price
  • 10127-400
    VWR® 12-Pk recycling option
    12/Pack
    Log in for availability
    Pack of 12 - $676.78