LPG Sample Cylinders, Koehler
300 ml
Catalog # 37000-218
Supplier: Koehler
Supplier #: K27852
Jump to:
Product Details & Documents
Welded stainless steel cylinders obtain representative samples of liquefied petroleum gases (LPG)
Two-valve type instruments feature a 20% outage tube and built-in pressure relief valve preset between 540 and 600psi (38 and 42kg/cm2).
Certifications: Conform to specifications: ASTM D1265, GPA 2140.
Specifications
- Capacity:300 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



