Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
LPG Sample Cylinders, Koehler

LPG Sample Cylinders, Koehler

150 ml
Catalog # 37000-216
Supplier:   Koehler
Supplier #:  K27851
LPG Sample Cylinders, Koehler
Catalog # 37000-216
Supplier:  Koehler
LPG Sample Cylinders, Koehler
LPG Sample Cylinders, Koehler

150 ml

Price
$3,193.57
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Welded stainless steel cylinders obtain representative samples of liquefied petroleum gases (LPG)

Two-valve type instruments feature a 20% outage tube and built-in pressure relief valve preset between 540 and 600psi (38 and 42kg/cm2).

Certifications: Conform to specifications: ASTM D1265, GPA 2140.

Specifications
  • Capacity:
    150 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.