VK Sponge, Contec®
VK Sponge, Single-Ply, Sterile, 12.7×12.7×1.5 cm (5×5×0.6")
Supplier: CONTEC, INC. MX
Supplier #: VKST0003
Jump to:
Product Details & Documents
The VertiKlean® Sponge is a disposable cleanroom sponge that features high sorbency polyester foam laminated to a Polynit wiping material with excellent abrasion resistance.
- High sorbency polyester foam makes sponges very sorbent
- Textured polyester surface for more aggressive cleaning tasks
- The sponge is laser cut to completely seal the edges
Designed for general cleaning and wiping, they are ideal for use in the laboratory. Autoclavable or supplied as a sterile, ready-to-use product.
Packaging: Non sterile sponges are packaged 8/bag, 3 bags/inner bag, 4 inner bags/case. Sterile sponges are packaged 1/bag, 15 bags/inner bag, 8 inner bags/case.
Specifications
- Description:VK Sponge, Single-Ply, Sterile
- L×W×H:12.7×12.7×1.5 cm (5×5×0.6")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



