Orders LinkContactUsLinkComponent
Wringers, Contec®
Wringers, Contec®
Catalog # 33502-260
Wringers, Contec®
Catalog # 33502-260
Supplier:  
Wringers, Contec®
Wringers, Contec®
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
WRINGER SS FLAT MOP 20L CS1 Catalog #89198-850
Product 89198-850 is similar to 33502-260 but manufacturer and specifications may vary.
Product Details & Documents
A variety of wringers designed for use with curved and flat mops.
  • Stainless steel and polypropylene wringers for a variety of Contec® bucket systems
  • Can be used with curved, flat, and string mops
  • Fully autoclavable
These wringers fit on Contec® 3.9 gallon (15 l), 5 gallon (20 l) , and 6.5 gallon (25 l) bucket systems. All wringers are compatible with a wide variety of disinfectants.
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Replacement spring, electropolished stainless steel, for Downpress Flat Mop Wringer 2756
    Replacement spring, electropolished stainless steel, for Downpress Flat Mop Wringer 2756
    76519-522
    Log in for availability
    Each - $20.10