Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Elbows, Labconco

Elbows, Labconco

90° Elbows, 30.5 cm (12"), 61 cm (24"), 25 (in ft. of straight duct), 6.4 kg (14 lbs.)
Catalog # 30182-136
Supplier:   Labconco
Supplier #:  5602100
Elbows, Labconco
Catalog # 30182-136
Supplier:  Labconco
Elbows, Labconco
Elbows, Labconco

90° Elbows, 30.5 cm (12"), 61 cm (24"), 25 (in ft. of straight duct), 6.4 kg (14 lbs.)

Price
$1,160.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

PVC elbows, both 45° and 90°, are compatible with thermoplastic duct

Designed and engineered for quick installation and minimum pressure losses, elbows feature belled end connections to receive PVC duct directly.

Specifications
  • Ø:
    30.5 cm (12")
  • Description:
    90° Elbows
  • Height:
    61 cm (24")
  • Shipping weight:
    6.4 kg (14 lbs.)
  • Equivalent resistance:
    25 (in ft. of straight duct)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.