Orders LinkContactUsLinkComponent
Conductivity Solutions, YSI
1 quart, 1,000 µmhos/cm, 0.50% at 25 °C
  23202-220
 :   YSI
 :  065270
Conductivity Solutions, YSI
  23202-220
 :  YSI
 :  
Conductivity Solutions, YSI
Conductivity Solutions, YSI
1 quart, 1,000 µmhos/cm, 0.50% at 25 °C
 :  
$155.79
Each (1qt US)
Use YSI Conductivity Solutions to calibrate conductivity cell and meter together as a system for maximum measurement accuracy.
Solutions are standardized against NIST Standard Reference Material potassium chloride (KCl). Each bottle includes calibration instruments and temperature correction information. Packaged in glass quart and plastic pint bottles.
  • Size:
    1 quart
  • Accuracy:
    0.50% at 25 °C
  • Conductivity:
    1,000 µmhos/cm