Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Pre-Cleaned Cubitainer Containers

Pre-Cleaned Cubitainer Containers

Level III Containers, 0.9 L (30.4 oz.)
Catalog # 23050-324
Supplier #:  EPC3155
Pre-Cleaned Cubitainer Containers
Catalog # 23050-324
Pre-Cleaned Cubitainer Containers

Level III Containers, 0.9 L (30.4 oz.)

Price
$1,566.96
Log in for availability
Case of 128
Minimum order quantity is 1
Product Details & Documents

These collapsible LDPE plastic containers have polypropylene closures and are convenient for space-saving and storage

Containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody-sealed and lot number-labeled for traceability to the enclosed Statement of Compliance.

Cap size: 38-400.

Specifications
  • Capacity:
    0.9 L (30.4 oz.)
  • Description:
    Level III Containers
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.