Orders LinkContactUsLinkComponent
Pre-Cleaned Cubitainer Containers
Pre-Cleaned Cubitainer Containers
Level III Containers, 3.8 L (1 gal.)
Catalog # 23050-254
Supplier #:  EPC3020
Pre-Cleaned Cubitainer Containers
Catalog # 23050-254
Pre-Cleaned Cubitainer Containers
Level III Containers, 3.8 L (1 gal.)
Price
$183.60
Log in for availability
Case of 12
Product Details & Documents
These collapsible LDPE plastic containers have polypropylene closures and are convenient for space-saving and storage
Containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody-sealed and lot number-labeled for traceability to the enclosed Statement of Compliance.
Cap size: 38-400.
Specifications
  • Capacity:
    3.8 L (1 gal.)
  • Description:
    Level III Containers