Orders LinkContactUsLinkComponent
Multi-Purpose Brushes, Perfex
Multi-Purpose Brushes, Perfex
Brush Head, Polyester, Green, 10"
Catalog # 22940-284
Supplier:   Perfex
Supplier #:  3040G
Multi-Purpose Brushes, Perfex
Catalog # 22940-284
Supplier:  Perfex
Multi-Purpose Brushes, Perfex
Multi-Purpose Brushes, Perfex
Brush Head, Polyester, Green, 10"
Price
$107.80
Log in for availability
Each
Product Details & Documents
These multi-purpose brushes contain moderately stiff polyester fiber.
  • Surround-set and flared fiber maintains surface contact at any angle and pprevents brush block from damaging surfaces
  • Durable fiber maintains shape and is visually coded to isolate areas of use
  • Impact-resistant polymer brush block withstands soaking and will not rot, decay, chip or crack
  • Non-conductive with superb abrasion and chemical resistance
  • Unaffected by water, grease, petroleum products, detergents, sanitizer, and solvents
Excellent for scrubbing irregular surfaces, as well as floors, walls, and ceilings.
Ordering information: Brush heads and handle sold separately.
Specifications
  • Bristle color:
    Green
  • Bristle material:
    Polyester
  • Description:
    Brush Head
  • Total length:
    10"
Accessories
Product Information
  • Catalog #
    Handle material
    Handle color
    Availability
    Price
  • Adjustable Handle with Short Connector
    Adjustable Handle with Short Connector
    53200-102
    Polymer
    White
    Log in for availability
    Each - $94.38