Orders LinkContactUsLinkComponent
Triangles
Triangles
Triangles, iron wire, 6.4 cm (2.5")
Catalog # 21750-096
Supplier #:  H-23870
Triangles
Catalog # 21750-096
Triangles
Triangles
Triangles, iron wire, 6.4 cm (2.5")
Price
$6.45
Log in for availability
Pack of 10
Product Details & Documents
These triangles are made up of iron wire (16 gauge) galvanized with plain clay pipe stems.
Smooth pipe stem triangles protect glassware on hotplates and over burner flames.
Specifications
  • Description:
    Triangles, iron wire, 6.4 cm (2.5")