Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Triangles

Triangles

Triangles, iron wire, 5.1 cm (2")
Catalog # 21750-094
Triangles
Catalog # 21750-094
Supplier:  
Triangles
Triangles

Triangles, iron wire, 5.1 cm (2")

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
TRIANGLE IRON WIRE CLAY 2IN (51MM) Catalog #21750-095

Product 21750-095 is similar to 21750-094 but manufacturer and specifications may vary.

Product Details & Documents

These triangles are made up of iron wire (16 gauge) galvanized with plain clay pipe stems.

Smooth pipe stem triangles protect glassware on hotplates and over burner flames.

Specifications
  • Description:
    Triangles, iron wire, 5.1 cm (2")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.