Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PH/ION METER 7350 W/STAN120V
Catalog # 16467-086
Supplier:
WTW
Expand image
PH/ION METER 7350 W/STAN120V
Catalog # 16467-086
Supplier:
WTW
Supplier Number:
50475738