$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
PH ELECTRODE,PREM BULB GLS RFL
16467-076
:
WTW
PH ELECTRODE,PREM BULB GLS RFL
16467-076
:
WTW
:
50476296