Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PYREX® Solution Bottles, Graduated, Corning

PYREX® Solution Bottles, Graduated, Corning

With Graduations, 19 L (5 gal.)
Catalog # 16145-161
Supplier:   Corning
Supplier #:  1596-19L
PYREX® Solution Bottles, Graduated, Corning
Catalog # 16145-161
Supplier:  Corning
PYREX® Solution Bottles, Graduated, Corning
PYREX® Solution Bottles, Graduated, Corning

With Graduations, 19 L (5 gal.)

Price
$790.00
Log in for availability
Case of 1
Product Details & Documents

PYREX® borosilicate glass.

  • With double scale (liters and gallons) black enamel graduations
  • Graduated in 500 ml increments
  • Will withstand hot air (dry) or steam (wet) sterilization
  • Necks are tooled to increase mechanical strength and for uniform stopper fit

Solution bottles are designed for storage of solutions and are ideal for media preparation.

Specifications
  • Capacity:
    19 L (5 gal.)
  • Description:
    With Graduations
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.