Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PYREX® Solution Bottles, Corning

PYREX® Solution Bottles, Corning

Plain, 45.5 L (12 gal.)
Catalog # 16145-080
Supplier:   Corning
Supplier #:  1595-12
PYREX® Solution Bottles, Corning
Catalog # 16145-080
Supplier:  Corning
PYREX® Solution Bottles, Corning
PYREX® Solution Bottles, Corning

Plain, 45.5 L (12 gal.)

Price
$1,650.00
Log in for availability
Case of 1
Product Details & Documents

Designed for storage of solutions or media preparation. The 13.25 L sizes are conventional shape; the 19 and 45.5 L sizes are similar to carboys in design. Necks are tooled to increase mechanical strength and for uniform stopper fit. Accepts No. 12 rubber stopper. Type I glass.

Available plain or with graduations. Graduated bottles have black enamel graduations in double scale, graduated in 500 ml increments.

 : Stopper not included.

 

Specifications
  • Capacity:
    45.5 L (12 gal.)
  • Description:
    Plain