Polypropylene Screw Cap, Wheaton
20-400, Blue
15507-140
: 240706-04
Unlined for use with WHEATON Glass Diagnostic Bottles.
- Solid or open top screw caps
- Use with thin-flange lyophilization stoppers sold separately
- Five colors available
- Autoclavable
- Cap Size:20-400
- Color:Blue