Polypropylene Screw Cap, Wheaton
20-400, White
Supplier: DWK Life Sciences (KIMBLE)
: 240706-02
Jump to:
Product Details & Documents
Unlined for use with WHEATON Glass Diagnostic Bottles.
- Solid or open top screw caps
- Use with thin-flange lyophilization stoppers sold separately
- Five colors available
- Autoclavable
Specifications
- Cap Size:20-400
- Color:White