Polypropylene Screw Cap, Wheaton
20-400, Yellow
Catalog # 15507-132
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 240716-05
Jump to:
Product Details & Documents
Unlined for use with WHEATON Glass Diagnostic Bottles.
- Solid or open top screw caps
- Use with thin-flange lyophilization stoppers sold separately
- Five colors available
- Autoclavable
Documents
Certificates
Search by Lot Number15507-124_0000034416 15507-124_0000047420 15507-134_0000006388 15507-134_0000036972 15507-136_0000037027
More
Wheaton Vial GUide
Product Literature
Specifications
- Cap Size:20-400
- Color:Yellow