Orders LinkContactUsLinkComponent
700 System Premium Receiver Head Assemblies, Techcon
700 System Premium Receiver Head Assemblies, Techcon
0.9 m (3")
Catalog # 14219-728
Supplier:   Techcon Systems
Supplier #:  70303RHB
700 System Premium Receiver Head Assemblies, Techcon
Catalog # 14219-728
Supplier:  Techcon Systems
700 System Premium Receiver Head Assemblies, Techcon
700 System Premium Receiver Head Assemblies, Techcon
0.9 m (3")
Price
$82.05
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Head assemblies ensure accident-proof dispensing and provide a secure and reliable link between the dispensing barrel and the controller
Complete dispensing control. Compatible with all industry-standard dispensing barrels. Designed to exact industry standards.
Specifications
  • Tubing length:
    0.9 m (3")