Orders LinkContactUsLinkComponent
700 System Premium Receiver Head Assemblies, Techcon
1.8 m (6")
  14219-720
 :  70506RHB
700 System Premium Receiver Head Assemblies, Techcon
  14219-720
700 System Premium Receiver Head Assemblies, Techcon
700 System Premium Receiver Head Assemblies, Techcon
1.8 m (6")
$80.35
Each
Head assemblies ensure accident-proof dispensing and provide a secure and reliable link between the dispensing barrel and the controller
Complete dispensing control. Compatible with all industry-standard dispensing barrels. Designed to exact industry standards.
  • Tubing length:
    1.8 m (6")