Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Project Timer, Sper Scientific

Project Timer, Sper Scientific

Project timer, 130×64×20 mm (5×2⅞×⅞"), 71 g (2.5 oz.)
Catalog # 14003-158
Supplier:   Sper Scientific
Supplier #:  810017
Project Timer, Sper Scientific
Catalog # 14003-158
Supplier:  Sper Scientific
Project Timer, Sper Scientific
Project Timer, Sper Scientific

Project timer, 130×64×20 mm (5×2⅞×⅞"), 71 g (2.5 oz.)

Price
$46.82
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

This long range digital countdown project timer allows you to set a target day for completion of a project up to 8 years in the future and the unique timer counts down to your deadline in days, hours, minutes, seconds and ¹/₁₀₀ seconds on a 12 digit LCD.

  • Standard clock, timer, and stopwatch modes
  • A tripod back enables the project timer to stand on a bench top

View either the target date or the actual count down. You can insert the name of your project into a special window which can be seen on the timer face.

Certifications: N.I.S.T. traceable certificate of compliance for 14003-160.

Ordering information: One AAA battery included.

Specifications
  • Description:
    Project timer
  • W×H×D:
    130×64×20 mm (5×2⅞×⅞")
  • Weight:
    71 g (2.5 oz.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.