Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

VWR® Modular Heating Sand Baths

Sand Bath for 2 Block Heater, Double, 9.5 x 14.9 x 6.4 cm (33/4 x 57/8 x 21/2")
  13259-264
VWR® Modular Heating Sand Baths
  13259-264
VWR® Modular Heating Sand Baths
VWR® Modular Heating Sand Baths

Sand Bath for 2 Block Heater, Double, 9.5 x 14.9 x 6.4 cm (33/4 x 57/8 x 21/2")

 
$644.94
   
Each
 

For use in heating blocks, these sand baths are ideal for irregular vessels.

  • Now with Best-in-Class, Exclusive 5-Year Warranty

Stainless steel construction for superior corrosion resistance. Designed to hold sand, shot, or nonvolatile liquids. Right angle thermometers (61220-740 and 61220-742) have an accuracy rating of ± 1°C.

 
  • Description:
    Sand Bath for 2 Block Heater
  • L×W×H:
    9.5 x 14.9 x 6.4 cm (33/4 x 57/8 x 21/2")
  • Block size:
    Double
Accessories

 

  •  
    Description
     
     
  • 13259-272
    Sand for Sand Bath, 454 g (1 lb.)
       
    Each - $173.66

 

  •  
    Description
     
     
  • 13259-274
    Stainless Steel Shot for Sand Bath, 454 g (1 lb.)
       
    Each - $203.95

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.