VWR® Modular Heating Blocks for Vials
12 mm Vial Block, Vials, Ext.Ø 12 mm, 9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2"), 20
Catalog # 12621-124
Supplier: VWR International
Jump to:
Product Details & Documents
Designed for sample/serum and scintillation vials, these units are constructed from a solid anodized aluminum block.
- Now with Best-in-Class, Exclusive 5-Year Warranty
The close contact of tubes to block walls allows for maximum heat retention. Single block size.
Specifications
- Description:12 mm Vial Block
- For vials:Ext.Ø 12 mm
- L×W×H:9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2")
- For use with:Vials
- No. of wells:20



