Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Low Density Flat Poly Liners, Associated Bag

Low Density Flat Poly Liners, Associated Bag

Yellow, 40 gal. cans (liner circumference 72"), 36×48", 4 mil.
Catalog # 11215-920
Supplier:   Associated Bag
Supplier #:  71-6-37Y
Low Density Flat Poly Liners, Associated Bag
Catalog # 11215-920
Supplier:  Associated Bag
Low Density Flat Poly Liners, Associated Bag
Low Density Flat Poly Liners, Associated Bag

Yellow, 40 gal. cans (liner circumference 72"), 36×48", 4 mil.

Price
$161.55
Log in for availability
Case of 50
Product Details & Documents

Sort and identify different types of waste.

Strong, bottom-sealed liners are free of heavy metals, making them safe for landfill or incineration.

Specifications
  • Color:
    Yellow
  • Thickness:
    4 mil.
  • W×L:
    36×48"
  • Fits:
    40 gal. cans (liner circumference 72")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.