Low Density Flat Poly Liners, Associated Bag
Yellow, Step-on cans (liner circumference 36"), 18×30 ", 4 mil.
11215-914
: 71-6-31Y
Sort and identify different types of waste.
Strong, bottom-sealed liners are free of heavy metals, making them safe for landfill or incineration.
- Color:Yellow
- Thickness:4 mil.
- W×L:18×30 "
- Fits:Step-on cans (liner circumference 36")



