Orders LinkContactUsLinkComponent
Accessories for SharpVue Microscopes
Accessories for SharpVue Microscopes
Lens for Sharp Vue 4D; Magnification: 2 - 52× Optical/up to 580× digital
Catalog # 78620-378
New Product
Supplier:   Aven Tools
Supplier #:  26700-138-4D
Accessories for SharpVue Microscopes
Catalog # 78620-378
Supplier:  Aven Tools
New Product
Accessories for SharpVue Microscopes
Lens for Sharp Vue 4D; Magnification: 2 - 52× Optical/up to 580× digital
Price
$366.42
Log in for availability
Each
Product Details & Documents
Specifications
  • Description:
    Lens for Sharp Vue 4D; Magnification: 2 - 52× Optical/up to 580× digital