Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Basic Vortex Mixer, Thermo Scientific

Basic Vortex Mixer, Thermo Scientific

Basic vortex mixer (Black Friday version), 100/240 V, 50/60 Hz, 100 – 240 V, Up to 19 tubes, 290×270×240 mm (11.4×10.7×9.4"), 200 - 3000 rpm, Touch, continous, 6.5 kg, 6.8 kg (14.99 lbs.), 0.5 kg
Catalog # BF77041-354
New Product
Supplier #:  88882011
Basic Vortex Mixer, Thermo Scientific
Catalog # BF77041-354
Supplier:  
New Product
Basic Vortex Mixer, Thermo Scientific
Basic Vortex Mixer, Thermo Scientific

Basic vortex mixer (Black Friday version), 100/240 V, 50/60 Hz, 100 – 240 V, Up to 19 tubes, 290×270×240 mm (11.4×10.7×9.4"), 200 - 3000 rpm, Touch, continous, 6.5 kg, 6.8 kg (14.99 lbs.), 0.5 kg

Product Details & Documents

Thermo Scientific™ Basic Vortex Mixer is ideal for quick, uniform mixing of samples with precise speed control.

  • Digital speed display
  • Continuous and touch modes
  • Speed range: 200 to 3000 rpm
  • Speed accuracy: ±5% rpm
  • Convenient speed adjustments by knob control
  • Stable for long-term operations
  • Multiple accessories for different vessels

Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.

Motor type: DC motor; Orbit: 4 mm; Material: ABS housing

Certifications: cCSAus.

Ordering information: 2 years of warranty.

Specifications
  • Capacity:
    Up to 19 tubes
  • Description:
    Basic vortex mixer (Black Friday version)
  • L×W×H:
    290×270×240 mm (11.4×10.7×9.4")
  • Electrical:
    100/240 V, 50/60 Hz
  • Shipping weight:
    6.8 kg (14.99 lbs.)
  • Control type:
    Touch, continous
  • Speed range:
    200 - 3000 rpm
  • Max. load:
    0.5 kg
  • Weight:
    6.5 kg
  • Voltage:
    100 – 240 V
  • Cat. No.:
    BF77041-354
  • Orbit:
    4 mm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Rubber bands, 6 per pack
    Rubber bands, 6 per pack
    76417-548
    Log in for availability
    Each - $18.61

Product Information

  • Catalog #
    Availability
    Price
  • Multi-power adaptor, 220 V, EU/UK/CHN plug
    Multi-power adaptor, 220 V, EU/UK/CHN plug
    76417-594
    Log in for availability
    Each - $77.34

Product Information

  • Catalog #
    Availability
    Price
  • Power adaptor, 120 V, US plug
    Power adaptor, 120 V, US plug
    76417-596
    Log in for availability
    Each - $56.79

Product Information

  • Catalog #
    For use with
    Availability
    Price
  • Platform for flat bottomed vessels, 87.5×10 mm
    Platform for flat bottomed vessels, 87.5×10 mm
    76417-608
    Digital vortex mixer
    Log in for availability
    Each - $33.59

Product Information

  • Catalog #
    For use with
    Availability
    Price
  • Tube holder, 10 mm Ø×19
    Tube holder, 10 mm Ø×19
    76417-612
    Digital vortex mixer
    Log in for availability
    Each - $46.34

Product Information

  • Catalog #
    For use with
    Availability
    Price
  • Microplate tray, black, 136.1×97.3×15 mm
    Microplate tray, black, 136.1×97.3×15 mm
    76417-610
    Digital vortex mixer
    Log in for availability
    Each - $33.59

Product Information

  • Catalog #
    For use with
    Availability
    Price
  • Tube holder, 12 mm Ø×13
    Tube holder, 12 mm Ø×13
    76417-614
    Digital vortex mixer
    Log in for availability
    Each - $46.34

Product Information

  • Catalog #
    For use with
    Availability
    Price
  • Tube holder, 15 mm Ø×12
    Tube holder, 15 mm Ø×12
    76417-616
    Digital vortex mixer
    Log in for availability
    Each - $46.34

Product Information

  • Catalog #
    For use with
    Availability
    Price
  • Tube holder, 20 mm Ø×7
    Tube holder, 20 mm Ø×7
    76417-618
    Digital vortex mixer
    Log in for availability
    Each - $46.34

Product Information

  • Catalog #
    For use with
    Availability
    Price
  • Tube holder, 26mm Ø×4
    Tube holder, 26mm Ø×4
    76417-620
    Digital vortex mixer
    Log in for availability
    Each - $46.34

Product Information

  • Catalog #
    For use with
    Availability
    Price
  • Rubber platform, black, 82×16.5 mm
    Rubber platform, black, 82×16.5 mm
    76417-606
    Digital vortex mixer
    Log in for availability
    Each - $33.59

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.