Tapestri® Software
Tapestri® Single-Cell Multiple Myeloma Software, portal access, Annual access to Tapestri® Single-Cell Multiple Myeloma analysis software hosted on portal.
Catalog # 78542-224
New Product
Supplier: MISSION BIO, INC.
Supplier #: MB06-0009
Jump to:
Product Details & Documents
Access to the cloud analysis pipeline for data processing is included. For additional tertiary analysis and specialized reporting, we offer user friendly software. Pipelines specialized in the analysis of Genome Editing, Multiple Myeloma, and Genome Integrity.
Ordering information: Does not include instrument, reagents or materials for processing samples. Access is provided to all laboratory users and starts on the order date.
Specifications
- Description:Tapestri® Single-Cell Multiple Myeloma Software, portal access
- Includes:Annual access to Tapestri® Single-Cell Multiple Myeloma analysis software hosted on portal.