VWR® Fluorometer
Product Details & Documents
VWR® Fluorometer for quantitation of DNA, RNA, and Proteins. Compact with touch screen control panel. Compatible with VWR® quantitation kits and kits from leading suppliers.
Sensitive quantitation down to 0.1 pg/µl ds DNA
- Large color touch screen for programming and operation
- Two models for single or eight-sample processing
- Built-in reagent calculators for sample preparation
- Compact and portable (11×6" footprint)
Sample volume: 1 to 20 µl
Compatible sample tubes: 0.5 ml transparent thin wall PCR tube
Linearity range: Five orders of magnitude
Sensitivity: dsDNA: 0.01 ng/µl, ssDNA: 0.05 ng/µl, RNA: 0.25 ng/µl, microRNA: 0.05 ng/µl
Calibration: 2 or 3 points
Control panel: 5" color touch screen
Working environment: 15 to 30 ℃, <75% RH, indoor use
Certifications: Meets ASTM standards. EN-342.
Accessories information: Accessories not included.
Documents
VWR® Fluorometer Brochure.pdf
Product Literature
Specifications
- Wavelength:Excitation: Blue: 460 - 480 nm, Red: 630 - 650 nm; Emission: Green: 500 - 535 nm, Red: 670 - 710 nm
- Description:VWR® Fluorometer, 8-channel, 115 - 240 V
- Cat. No.:VWRI210-000173
- Detector:Silicon photomultiplier
- Scanning speed:≤5 s/sample
- W×D×H:10.8×24.1×5.7 cm (4.25×9.5×2.25")
- Sample handling:8 (sample throughput)
- Electrical:100 - 240 VAC
- Light Source:LED
- Weight:0.5 kg (1.1 lbs.)
Accessories
Product Information
- Catalog #AvailabilityPrice
- VWR® DS-DNA HS quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® 1× ready-to-use DS-DNA hs quant kit, 100 assaysVWR® 1× ready-to-use DS-DNA hs quant kit, 100 assaysVWRI210-000160Log in for availabilityEach - $135.20
Product Information
- Catalog #AvailabilityPrice
- VWR® DS-DNA BR quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® 1× ready-to-use DS-DNA BR quant kit, 100 assaysVWR® 1× ready-to-use DS-DNA BR quant kit, 100 assaysVWRI210-000162Log in for availabilityEach - $135.20
Product Information
- Catalog #AvailabilityPrice
- VWR® SS-DNA quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® RNA HS quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® RNA BR quant kit, 100 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® fluorometer tubes 0.5 ml, 250 per packVWR® fluorometer tubes 0.5 ml, 250 per packVWRI210-000166Log in for availabilityPack of 250 - $72.80
Product Information
- Catalog #AvailabilityPrice
- Fluorometer tubes, 0.2 ml, pk/1000
Product Information
- Catalog #AvailabilityPrice
- Fluorometer tube-strips, 0.2 ml, pk/120
Product Information
- Catalog #AvailabilityPrice
- VWR® DS-DNA HS quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® 1× DS-DNA HS quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® DS-DNA BR quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® 1× DS-DNA BR quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® SS-DNA quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® RNA HS quant kit, 500 assays
Product Information
- Catalog #AvailabilityPrice
- VWR® RNA BR quant kit, 500 assays
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



