Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Column Component Closures

Column Component Closures

Column component closure, with one ¹/₄" FNPT port, 58 mm
Catalog # 78473-816
New Product
Supplier:   SAVILLEX
Supplier #:  600-058-17
Column Component Closures
Catalog # 78473-816
Supplier:  SAVILLEX
New Product
Column Component Closures

Column component closure, with one ¹/₄" FNPT port, 58 mm

Price
$64.89
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.

Specifications
  • Description:
    Column component closure, with one ¹/₄" FNPT port, 58 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.