Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Microbial Count Plate

Psychrophilic Bacteria count plate, Red colony, 36±1 °C, 8-18h incubation
Catalog # 78172-248
New Product
 :  MCPG02925
Microbial Count Plate
Catalog # 78172-248
New Product
CAS Number:  
Microbial Count Plate

Psychrophilic Bacteria count plate, Red colony, 36±1 °C, 8-18h incubation

CAS Number:  
Price
$243.00
Log in for availability
Pack of 25
Product Details & Documents

GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.

  • Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required

GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.

Specifications

Specifications

  • Description:
    Psychrophilic Bacteria count plate
  • Format:
    Red colony, 36±1 °C, 8-18h incubation

Specifications