Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Microbial Count Plate

Geobacillus stearothermophilus count plate, Red colony, 36±1 °C, 18-24h incubation
Catalog # 78172-722
New Product
 :  MCPG01725
Microbial Count Plate
Catalog # 78172-722
New Product
CAS Number:  
Microbial Count Plate

Geobacillus stearothermophilus count plate, Red colony, 36±1 °C, 18-24h incubation

CAS Number:  
Price
$243.00
Log in for availability
Pack of 25
Product Details & Documents

GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.

  • Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required

GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.

Specifications

Specifications

  • Description:
    Geobacillus stearothermophilus count plate
  • Format:
    Red colony, 36±1 °C, 18-24h incubation
  • Cat. No.:
    78172-722

Specifications