Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Microbial Count Plate

Staph count plate, Red colony, 36±1 °C, 48±2h incubation
Catalog # 78173-198
New Product
 :  MCPG00225
Microbial Count Plate
Catalog # 78173-198
New Product
CAS Number:  
Microbial Count Plate

Staph count plate, Red colony, 36±1 °C, 48±2h incubation

CAS Number:  
Price
$85.05
Log in for availability
Pack of 25
Product Details & Documents

GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.

  • Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required

GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.

Specifications

Specifications

  • Description:
    Staph count plate
  • Format:
    Red colony, 36±1 °C, 48±2h incubation

Specifications