Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Microbial Count Plate

Microbial Count Plate

Enterobacteriaceae EB count plate, Purple, red colony, 36±1 °C, 24±2h incubation
Catalog # 78173-170
New Product
Supplier #:  MCPG01625
Microbial Count Plate
Catalog # 78173-170
New Product
CAS Number:  
Microbial Count Plate

Enterobacteriaceae EB count plate, Purple, red colony, 36±1 °C, 24±2h incubation

CAS Number:  
Price
$121.50
Log in for availability
Pack of 25
Product Details & Documents

GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.

  • Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required

GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.

Specifications

Specifications

  • Description:
    Enterobacteriaceae EB count plate
  • Format:
    Purple, red colony, 36±1 °C, 24±2h incubation

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.