Microbial Count Plate
Shigella count plate, Red colony, , 36±1 °C, 24±2h incubation
Catalog # 78172-954
New Product
Supplier: GVS FILTER TECHNOLOGY
Supplier #: MCPG02525
Jump to:
Product Details & Documents
GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.
- Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required
GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.
Specifications
Specifications
- Description:Shigella count plate
- Format:Red colony, , 36±1 °C, 24±2h incubation
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



