Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Quick Clip

Quick Clip

Quick clip, White, Polyamide, 220 mm
Catalog # 78117-020
New Product
Supplier #:  8219A-220
Quick Clip
Catalog # 78117-020
New Product
Quick Clip
Quick Clip

Quick clip, White, Polyamide, 220 mm

Price
$868.54
Log in for availability
Case of 100
Product Details & Documents

Quick clip provides an instant seal for bags.

  • Provides an instant powder- and water-tight seal for bags

Quick clips can be used over and over again, providing a convenient and cost-eefective solution to sealing different types of bags.

Specifications
  • Color:
    White
  • Description:
    Quick clip
  • Length:
    220 mm
  • Material:
    Polyamide