0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER LOOP AS-AP AUTOSAMPLER 1.2ML
Catalog # MSPP-074998
Supplier:
THERMO ELECTRON NORTH AMERICA MS
New Product
Expand image
BUFFER LOOP AS-AP AUTOSAMPLER 1.2ML
Catalog # MSPP-074998
Supplier:
THERMO ELECTRON NORTH AMERICA MS
Supplier Number:
074998
New Product