Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Modified Nutrient Broth

Modified Nutrient Broth

Modified nutrient broth, 500 ml, Liquid, 730 days
Catalog # 77442-468
Supplier:   Teknova
Supplier #:  N1275
Modified Nutrient Broth
Catalog # 77442-468
Supplier:  Teknova
CAS Number:  
Modified Nutrient Broth

Modified nutrient broth, 500 ml, Liquid, 730 days

CAS Number:  
Price
$71.62
Log in for availability
Each
Minimum order quantity is 8
Product Details & Documents

Modified nutrient broth is a general-purpose liquid medium for cultivation and routine propagation of nonfastidious microorganisms. pH: 6.80 ±0.20.

  • General bacterial cultivation: Supports robust growth of nonfastidious organisms such as Escherichia coliStaphylococcus aureus, and Bacillus subtilis for routine culture and maintenance

    Inoculum preparation: Used to prepare standardised inoculum for downstream biochemical identification and antimicrobial susceptibility testing

    Enumeration and sampling: Employed for microbial counts in food, water, and environmental samples via serial dilution and recovery

    Sterility testing: Serves as a nonselective medium to detect microbial contamination in pharmaceuticals, media, and process controls

    Recovery of stressed cells: Facilitates resuscitation of sublethally injured bacteria following heat, desiccation, or sanitiser exposure

    QC and method validation: Provides a consistent growth matrix for positive controls and assay verification with strains like Pseudomonas aeruginosa and Salmonella enterica

    Teaching and training: Ideal for microbiology education labs to demonstrate microbial growth, turbidity, and aseptic technique

    Starter cultures for downstream workflows: Generates healthy log-phase cultures for cloning, plasmid prep, and protein expression pre-cultures

Beef extract and pancreatic digest of casein supply peptides, amino acids, and growth factors that support the culture of a broad range of bacteria, including E. coli and Staphylococcus aureus. It is suitable for inoculum preparation, maintenance of laboratory strains, and microbial enumeration in quality-control workflows.

  • Recovery of Stressed Cells: Facilitates resuscitation of sublethally injured bacteria following heat, desiccation, or sanitizer exposure.
  • QC and Method Validation: Provides a consistent growth matrix for positive controls and assay verification with strains like Pseudomonas aeruginosa and Salmonella enterica.
  • Teaching and Training: Ideal for microbiology education labs to demonstrate microbial growth, turbidity, and aseptic technique.
  • Starter Cultures for Downstream Workflows: Generates healthy log-phase cultures for cloning, plasmid prep, and protein expression pre-cultures.

Specifications

Specifications

  • Media Format:
    Liquid
  • Description:
    Modified nutrient broth
  • Fill volume:
    500 ml
  • Shelf life:
    730 days
  • Storage Conditions:
    RT
  • Cat. No.:
    77442-468
  • Formulation:
    0.3% Beef extract; 0.5% Tryptone
  • Sterility:
    Sterile

Specifications