Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

55C MMLV-RT

New Product
55C MMLV-RT
New Product
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A highly robust, thermostable reverse transcriptase, ideal for cDNA synthesis from RNA with a high secondary structure such as viral RNA genomes.

  • Thermostable reverse transcriptase activity up to 60 °C
  • Ideal for cDNA synthesis from RNA with high secondary structure such as viral RNA genomes
  • Sensitive detection of low copy number RNA targets
  • Perfect for developing fast, highly reproducible RT-qPCR assays

For cDNA synthesis, a higher reaction temperature is sometimes desirable as it reduces RNA secondary structures which can inhibit reverse transcription and it minimizes nonspecific primer binding. 55C MMLV-RT is a highly thermostable reverse transcriptase with reduced RNase H activity, that allows very efficient first-strand cDNA synthesis at temperatures up to 60 °C. This improves the cDNA yield from difficult RNA targets, such as viral RNA genomes, that require higher temperature to denature strong RNA secondary structure.

 : ISO 13485

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 78577-120
  • 78577-106
  • Format:
  • Enzyme with/without buffer
  • Enzyme with/without buffer
  • Features:
  • Glycerol free
  • Glycerol free
  • Hot start:
  • No
  • No
  • High fidelity:
  • No
  • No
  • Amplicon Size:
  • ≤12 kb
  • ≤12 kb
  • Processivity:
  • Fast
  • Fast
  • Product overhang:
  • No
  • No
  • Storage Conditions:
  • –20 °C
  • –20 °C
  • Application:
  • Inhibitor rich sample
  • Inhibitor rich sample

Specifications

Product Family Options

Product Information

  • Concentration
    Reaction volume
    Units
    Volume
    No. of reactions
    Size
    Availability
    Price
  • 78577-120
    55C MMLV-RT
    55C MMLV-RT
    200 U/µl
    20 µl
    10000
    50 µl
    >700
    1×50 µl
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $256.00
    $256.00
    Each(1/Itm/Bx)
       $256.00
     

    Supplier Number: MDX117-005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78577-120
    Format
    Enzyme with/without buffer
    Features
    Glycerol free
    Hot start
    No
    High fidelity
    No
    Amplicon Size
    ≤12 kb
    Processivity
    Fast
    Product overhang
    No
    Storage Conditions
    –20 °C
    Application
    Inhibitor rich sample
    New Product
  • 78577-106
    55C MMLV-RT
    55C MMLV-RT
    200 U/µl
    20 µl
    40000
    200 µl
    >2800
    1×200 µl
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $735.00
    $735.00
    Each(1/Itm/Bx)
       $735.00
     

    Supplier Number: MDX117-02
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78577-106
    Format
    Enzyme with/without buffer
    Features
    Glycerol free
    Hot start
    No
    High fidelity
    No
    Amplicon Size
    ≤12 kb
    Processivity
    Fast
    Product overhang
    No
    Storage Conditions
    –20 °C
    Application
    Inhibitor rich sample
    New Product