Orders LinkContactUsLinkComponent
Field Wire Input Adapter
Field Wire Input Adapter
Projected signs field sire input adapter, black
Catalog # 78663-696
New Product
Supplier:   Brady Worldwide
Supplier #:  178983
Field Wire Input Adapter
Catalog # 78663-696
Supplier:  Brady Worldwide
New Product
Field Wire Input Adapter
Field Wire Input Adapter
Projected signs field sire input adapter, black
Price
$83.99
Log in for availability
Each
Product Details & Documents
Connect external control systems to any Brady LED projector without cutting or damaging the factory wiring.
  • PLC-ready integration
  • Tool-free connection
  • Fully compatible
  • Supports DC input signals up to 10 V
The field wire input adapter provides seamless connection to Programmable logic controllers (PLCs) and other automated systems. Convenient screw terminals enable fast, secure connections. The adapter is designed for use with any Brady LED projector.
Specifications
  • Description:
    Projected signs field sire input adapter, black