Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Water

Glass bottle, 4 L
Catalog # 78606-597
New Product
Supplier:   MilliporeSigma
 :  667404-4X4L
Water
Catalog # 78606-597
Supplier:  MilliporeSigma
New Product
CAS Number:  
Water

Glass bottle, 4 L

CAS Number:  
Price
$650.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Hydrogen oxide
  • MDL Number:MFCD00011332
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.