Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Diffuser Attachment for Mighty Scope ClearVue Digital Microscope

Diffuser Attachment for Mighty Scope ClearVue Digital Microscope

Diffuser attachment for mighty Scope ClearVue, Plastic material,
Catalog # 78621-210
New Product
Supplier:   Aven Tools
Supplier #:  26700-220-DIFF
Diffuser Attachment for Mighty Scope ClearVue Digital Microscope
Catalog # 78621-210
Supplier:  Aven Tools
New Product
Diffuser Attachment for Mighty Scope ClearVue Digital Microscope

Diffuser attachment for mighty Scope ClearVue, Plastic material,

Price
$122.44
Log in for availability
Each
Product Details & Documents

Designed to be used with Aven's Mighty Scope ClearVue digital microscope. When in use, this attachment disperses the LED light from the Mighty Scope ClearVue for soft, even illumination. For best results while in use, the recommended working is between 2.5 to 3.5".

  • Custom made attachment for the Mighty Scope ClearVue Digital Microscope
  • Evenly disperses LED light from the Mighty Scope ClearVue for soft and even illumination
  • Eliminates glares and hot spots caused by bright lights
  • Ideal when examining shiny or reflective parts
  • Easy to install and remove
Specifications
  • Description:
    Diffuser attachment for mighty Scope ClearVue, Plastic material,
  • Working Distance:
    2.5 - 3.5" (when in use)
  • Cat. No.:
    78621-210
  • W×D×H:
    4×4.75×1.25"
  • Weight:
    0.05 lbs.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.