Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Probe Double Sided Scraper Bent Tips

Probe Double Sided Scraper Bent Tips

Probe double sided with bent flat and curved point, weight: 0.04 lbs, Stainless steel, 165 mm, 0.04 lbs.
Catalog # 78620-710
New Product
Supplier:   Aven Tools
Supplier #:  20035
Probe Double Sided Scraper Bent Tips
Catalog # 78620-710
Supplier:  Aven Tools
New Product
Probe Double Sided Scraper Bent Tips

Probe double sided with bent flat and curved point, weight: 0.04 lbs, Stainless steel, 165 mm, 0.04 lbs.

Price
$6.90
Log in for availability
Each
Product Details & Documents

With it's needle-sharp point, this stainless steel probe offers a variety of uses, including cold solder probing, hand inspection, micro positioning and lead adjusting.

  • Manufactured from 400 series stainless steel

  • Excellent corrosion resistance

    Polished

  • Sharp tip for precision work around delicate circuits and components
  • Double sided with bent flat and curved points
  • Eight sided handle is knurled at each end for extra grip

Specifications
  • Description:
    Probe double sided with bent flat and curved point, weight: 0.04 lbs
  • Length:
    165 mm
  • Material:
    Stainless steel
  • Weight:
    0.04 lbs.