Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
LED Ring Light, Pro 80

LED Ring Light, Pro 80

Pro Series 80 LED ring light, 100 - 240 V
Catalog # 78620-486
New Product
Supplier:   Aven Tools
Supplier #:  26200B-218
LED Ring Light, Pro 80
Catalog # 78620-486
Supplier:  Aven Tools
New Product
LED Ring Light, Pro 80

Pro Series 80 LED ring light, 100 - 240 V

Price
$1,870.88
Log in for availability
Each
Product Details & Documents

Pro Series 40 white (daylight) LED ring light with four quadrant sector and brightness controller and power supply.

  • Machined aluminum alloy housing for durability

    Two fully adjustable dimming controls

    360° shadow free illumination

    Fits lenses up to 66 mm (2.6")

    Blacklight (320 to 400 nm) does not represent a hazard under normal use conditions

Pro Series 80 dual output LED ring light features an outer ring of 40 white LEDs and an inner ring of 40 UV LEDs operating at 360 nm. These high quality LEDs are set at different angles to create uniform illumination across a wide range of working distances. The UV light is excellent for detecting fluoresces in adhesives, laminates and conformal coatings, forgery analysis, currency verification, gemstone and mineral inspection. Each ring and LED segment can be independently controlled using our advanced eight segment LED controller.

Dual dimming controls provide maximum color stability at all lighting levels. CNC machined from a solid block of aluminum alloy allowing optimal thermal management and performance in a rugged attractive package. Supplied with standard universal power source for worldwide operation.

LEDs: 80 LEDs (40 white and 40 UV)

Lighting control: Light control box with 8 quadrant switches and 2 dimmer controls

LED color temperature: White LED: 6000 K; UV LEDs: 360 nm

Inside ring diameter: 66.8 mm (2.63")

Outside ring diameter: 105 mm (4.11")

Height: 26 mm (1.02")

Cable length: 182.88 cm (6')

Min. working distance: 88.9 mm (3.5")

Ideal working distance: 114 mm (4.5')

Maximum working distance: 178 mm (7.0")

Control box dimensions: 135×100×30 mm

Ring light dimensions: 105×26 mm

Weight: Ring light: 0.55 lbs.; Control box: 0.2 lbs.

Delivery information: Package contains Ring light, control box, HDMI cable, power supply, safety glasses.

Specifications
  • Description:
    Pro Series 80 LED ring light
  • Electrical:
    100 - 240 V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.